Oxygen determination bottle, acc. to Winkler, normal capacity 100 - 150 ml, joint NS 14/23 with NS glass stopper, bottle and stopper adjusted and marked, soda lime glass, numbered from 700-919 1 * 1 items
Catalog # LENZ81004222
Supplier: Lenz Laborglas GmbH & CO.KG
Frequently Bought Together