Order Entry
Netherlands
ContactUsLinkComponent
Innovative detoxification buffer removes endotoxins and proteins from plasmid DNA for use in transfection. Replacement buffer solution for use with NucleoSpin Plasmid/96 Plasmid Transfection-grade kits. 1 * 1.000 mL
Catalog # MANA740495.1000
Supplier:  MACHEREY-NAGEL
Innovative detoxification buffer removes endotoxins and proteins from plasmid DNA for use in transfection. Replacement buffer solution for use with NucleoSpin Plasmid/96 Plasmid Transfection-grade kits. 1 * 1.000 mL
Catalog # MANA740495.1000
Supplier:  MACHEREY-NAGEL
Frequently Bought Together