Order Entry
Netherlands
ContactUsLinkComponent
Chemical protection glove, Winter-lined, Neoprene, Crinkled grip pattern, Cat. III, Black, Latex-free, Winter-lined, Withstands contact heat up to 500°C, Extra long, for heavy work 1 * 1 PAIR
Catalog # EJEN494-10
Supplier:  EJENDALS
Chemical protection glove, Winter-lined, Neoprene, Crinkled grip pattern, Cat. III, Black, Latex-free, Winter-lined, Withstands contact heat up to 500°C, Extra long, for heavy work 1 * 1 PAIR
Catalog # EJEN494-10
Supplier:  EJENDALS
Frequently Bought Together