Kemio benchtopkit 1 * 1 items
Catalog # PALIKEMR10DIS
Supplier: Palintest
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Accessories for Mettler balances
Supplier: Mettler - Toledo
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® pHenomenal® Electrode Stand
Supplier: VWR Collection
pH electrodes, SenTix® PLUS
Supplier: WTW
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Titration beakers
Supplier: Mettler - Toledo
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Wipes for delicate tasks, Kimtech™ Science
Supplier: KIMBERLY CLARK
Swab-sampler with buffered peptone water broth
Supplier: 3M Food Safety
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
Plastic cuvette for LICO
Supplier: Hach