Order Entry
Netherlands
ContactUsLinkComponent
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 1260 MPA.S (25.00C), RTM25 1 * 500 mL
Catalog # PARGRTM25
Supplier:  PARAGON SCIENTIFIC
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 1260 MPA.S (25.00C), RTM25 1 * 500 mL
Catalog # PARGRTM25
Supplier:  PARAGON SCIENTIFIC
Frequently Bought Together