Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VISCOSITY STANDARD, ROTATIONAL SILICONE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 10.12 MPA.S (25.00C), RT10 1 * 600 mL
Catalog # PARGVIS-RT10-600
Supplier:
PARAGON SCIENTIFIC
Certificates
Expand image
VISCOSITY STANDARD, ROTATIONAL SILICONE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 10.12 MPA.S (25.00C), RT10 1 * 600 mL
Catalog # PARGVIS-RT10-600
Supplier:
PARAGON SCIENTIFIC
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
VWR® ARISTAR®, Multi-Element Calibration Standard IV in dil. Nitric Acid for ICP
Supplier: VWR Chemicals
VWR®, Refractive index Certified Reference Material at 20, 25 and 30 °C
Supplier: VWR Chemicals
Ammonium standard solution, 1000 mg/L NH₄ in water (from NH₄Cl) Certipur® standard for Ion Chromatography, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Potassium hydrogen phthalate, Certipur® Reag. Ph. Eur., Reag. USP volumetric standard, secondary reference material for alkalimetry, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
pH Standard Solutions, IUPAC, Certified
Supplier: Hach