VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 2610 MPA.S (25 C) 1 * 500 mL
Catalog # PARGN1000
Supplier: PARAGON SCIENTIFIC
VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 2610 MPA.S (25 C) 1 * 500 mL
Catalog # PARGN1000
Supplier: PARAGON SCIENTIFIC
Frequently Bought Together
Calcium carbonate 98.5-100.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Bottles, wide neck, with screw cap, Nalgene®
Supplier: Thermo Fisher Scientific
Supelco® Organic Reference Standard, Chlorophyll a
Supplier: Merck
CEFAZOLIN SODIUM SALT 1 * 75 mg
Supplier: Merck
Isoxaflutole-diketonitrile 1 * 10 mg
Supplier: EHRENSTORFER
Arsenic standards for ICP, TraceCERT®, Supelco®
Supplier: Merck
ACETYLTHIAMINE HYDROCHLORIDE, THIAMINE I 1 * 100 mg
Supplier: Merck
DIISOPROPYL ETHER 1 * 3 items
Supplier: Merck
3-Ethylphenol
Supplier: EHRENSTORFER
Buffer Solutions
Supplier: Thermo Fisher Scientific
Supelco® TraceCERT® Organic Reference Standard, Isoflucypram
Supplier: Merck
IRINOTECAN HYDROCHLORIDE 1 * 200 mg
Supplier: Merck
Latanoprost Related Compounds, Reference Standard
Supplier: USP
Supelco® Organic Reference Standard, 3-Methylbutanol
Supplier: Merck
pH buffer solutions, referenced at 20 °C, traceable to SRM from NIST, Fluka™
Supplier: Honeywell Chemicals
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals