Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 202.6 MPA.S (25 C) 1 * 500 mL
Catalog # PARGN100
Supplier:
PARAGON SCIENTIFIC
Certificates
Afbeelding vergroten
VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 202.6 MPA.S (25 C) 1 * 500 mL
Catalog # PARGN100
Supplier:
PARAGON SCIENTIFIC
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Reagent Reservoirs
Supplier: VWR Collection
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals