Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VISCOSITY STANDARD, FLOW CUP, UKAS ISO 17025 AND ISO 17034 CERTIFIED, VISCOSITY NOMINAL 135.9 MPA.S (20 C) 1 * 500 mL
Catalog # PARGC60
Supplier:
PARAGON SCIENTIFIC
Expand image
VISCOSITY STANDARD, FLOW CUP, UKAS ISO 17025 AND ISO 17034 CERTIFIED, VISCOSITY NOMINAL 135.9 MPA.S (20 C) 1 * 500 mL
Catalog # PARGC60
Supplier:
PARAGON SCIENTIFIC
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Hydrochloric acid 2 mol/l (2 N), AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ