Order Entry
Netherlands
ContactUsLinkComponent
Sartorius Y-Connector RS232 9-pin (male) to 2x RS232 9-pin (female). To connect Sartorius laboratory balances of the product family Entris® in parallel to a foot switch and a printer, a barcode scanner or a second display. Length: approx. 1.5 m, Colo 1 * 1 items
Catalog # SARBYCC-D09M-2D09F
Supplier:  Sartorius Balances
Sartorius Y-Connector RS232 9-pin (male) to 2x RS232 9-pin (female). To connect Sartorius laboratory balances of the product family Entris® in parallel to a foot switch and a printer, a barcode scanner or a second display. Length: approx. 1.5 m, Colo 1 * 1 items
Catalog # SARBYCC-D09M-2D09F
Supplier:  Sartorius Balances
Frequently Bought Together