Cr 0.1mg/l Mn 1mg/l Fe 10mg/l Cu 1mg/l Mo 0.1mg/l Se 0.2mg/l Sn 0.4mg/l As 0.05mg/l Pb 0.05mg/l. Solvent: HNO3 2% Quantity: 1 x 250ml 1 * 1 Fl
Catalog # MSPPRM033369L25
Supplier: MARKET SOURCE PART PROCESS
Cr 0.1mg/l Mn 1mg/l Fe 10mg/l Cu 1mg/l Mo 0.1mg/l Se 0.2mg/l Sn 0.4mg/l As 0.05mg/l Pb 0.05mg/l. Solvent: HNO3 2% Quantity: 1 x 250ml 1 * 1 Fl
Catalog # MSPPRM033369L25
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
ep Dualfilter T.I.P.S.® Racks, Filter Pipette Tips
Supplier: EPPENDORF
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Separating funnels, Squibb
Supplier: Lenz Laborglas GmbH & CO.KG
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals