Order Entry
Netherlands
ContactUsLinkComponent
Cubis MCE balance with state of the art TFT touch screen operation with brilliant, readable, display but uncomplex, easy to operate user interface. Actual scale division (d) 10 mg, nominal maximum load (Max) 8200g. Standard version non-verified, all 1 * 1 items
Catalog # SARBMCE8202S2S00-0
Supplier:  Sartorius Balances
Cubis MCE balance with state of the art TFT touch screen operation with brilliant, readable, display but uncomplex, easy to operate user interface. Actual scale division (d) 10 mg, nominal maximum load (Max) 8200g. Standard version non-verified, all 1 * 1 items
Catalog # SARBMCE8202S2S00-0
Supplier:  Sartorius Balances
Frequently Bought Together