STORAGE BIN MADE OF POLYETHYLENE (COLOUR BLUE ) 1 * 10 ST
KAKA269786-49
Frequently Bought Together
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transferpipetten, Hoge Prestaties
: VWR Collection
VWR® NITRILE LIGHT, Nitrilhandschoenen
: VWR Collection
VWR®, Serologische pipetten, standaardreeks
: VWR Collection
Nitril handschoenen, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrilhandschoenen
: VWR Collection
VWR®, gezichtstissues
: VWR Collection
pH-teststrips, MQuant®, Supelco®
: Merck