Order Entry
Netherlands
ContactUsLinkComponent
NONIDET® P-40 SUBSTITUTE, 10% AQUEOUS SOLUTION CRIMP STYLE VIAL: IS A NON-IONIC DETERGENT THAT IS PROVIDED FOR RESEARCH PURPOSES OR AS A 10% PROTEOMIC GRADE SOLUTION. 1 * 50 mL
Catalog # GENODG501C
Supplier:  G-Biosciences
NONIDET® P-40 SUBSTITUTE, 10% AQUEOUS SOLUTION CRIMP STYLE VIAL: IS A NON-IONIC DETERGENT THAT IS PROVIDED FOR RESEARCH PURPOSES OR AS A 10% PROTEOMIC GRADE SOLUTION. 1 * 50 mL
Catalog # GENODG501C
Supplier:  G-Biosciences
Frequently Bought Together