MASSONS TRICHROME STAIN KIT BEGINS WITH THE PROCESS AFTER STAINING, COLLAGEN WILL BE COLOURED IN BLUE, MUSCLE FIBERS, CYTOPLASM AND KERATIN IN RED AND THE NUCLEI BLUE/BLACK. 1 * 125 mL
Catalog # GENOBAQ085
Supplier: G-Biosciences
Frequently Bought Together