Order Entry
Netherlands
ContactUsLinkComponent
Detect Mouse Cd5l with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernat es, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK1414
Supplier:  Boster Bio
Detect Mouse Cd5l with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernat es, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK1414
Supplier:  Boster Bio
Frequently Bought Together