[EN]TISSUE CULT PLATE 12/2ML FLAT BOTTOM 1 * 50 ST
SARS83.3921
: SARSTEDT
Frequently Bought Together
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
: VWR Collection
VWR® Media Bottles, Square, PETG
: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
: VWR Collection
Deepwell Plates 96-well, 1000 µl
: EPPENDORF
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
PCR plates, 96-well, Armadillo™
: Thermo Fisher Scientific
Petri dishes, Sterilin®
: STERLIN
50ml Tube Bioreactor PK 180 1 * 180 items
: GRANT INSTRUMENTS
Coloured cap inserts, Capinsert™
: Simport Scientific
Adhesive PCR film seals, QuickSeal PCR™
: iST Scientific Ltd
Storage tubes, MATRIX™
: THERMO MATRIX TECHNOLOGIES
Ice buckets with lids, Azlon®
: SciLabware
Containers with optional snap caps, Nunc™
: Thermo Fisher Scientific
TISSUE CULTURE PLATES 6-WELL FLAT BOTTO
: SARSTEDT
Microtubes, with thread
: SARSTEDT