Order Entry
Netherlands
ContactUsLinkComponent
Ground Joint glAdaptors, Black screw cap made of PPS with gl45 thread, movable insert with ground joint made of PTFE. Transition fro m a ground joint to a glass thread. The body can be turned independently from the screw cap. The completely assembled 1 * 1 items
Catalog # BOHLD740-40
Supplier:  Bohlender
undefined
Ground Joint glAdaptors, Black screw cap made of PPS with gl45 thread, movable insert with ground joint made of PTFE. Transition fro m a ground joint to a glass thread. The body can be turned independently from the screw cap. The completely assembled 1 * 1 items
Catalog # BOHLD740-40
Supplier:  Bohlender
Frequently Bought Together