Order Entry
Netherlands
ContactUsLinkComponent
Flat Flange Vessels, Made of transparent PFA model with flat bottom and pore-free surface. Flange with circular groove. Heatable wi th thermostats or in a heating hood. 1 * 1 items
Catalog # BOHLB325-01
Supplier:  Bohlender
undefined
Flat Flange Vessels, Made of transparent PFA model with flat bottom and pore-free surface. Flange with circular groove. Heatable wi th thermostats or in a heating hood. 1 * 1 items
Catalog # BOHLB325-01
Supplier:  Bohlender
Frequently Bought Together