Order Entry
Netherlands
ContactUsLinkComponent
RH506 - behrotest serial heating system for the reflux destillation of 500 ml round-bottom flasks for 6 sample positions 1 * 1 items
Catalog # BEHRB00602399
Supplier:  BEHR
RH506 - behrotest serial heating system for the reflux destillation of 500 ml round-bottom flasks for 6 sample positions 1 * 1 items
Catalog # BEHRB00602399
Supplier:  BEHR
Frequently Bought Together