Pasteur pipette, soda-lime glass, total length approx. 310 mm cap.approx. 1,5 ml 1 * 1.000 items
Catalog # BRND747730
Supplier: Brand
Pasteur pipette, soda-lime glass, total length approx. 310 mm cap.approx. 1,5 ml 1 * 1.000 items
Catalog # BRND747730
Supplier: Brand
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
Dialysis membrane tubing Spectra/Por® 6
Supplier: Spectrum Laboratories
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Pouring rings for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF