Vinyl Chloride Solution, 5000 µg/mL in Methanol 1 * 5 mL
Catalog # EHERGA09010500ME
Supplier: EHRENSTORFER
Vinyl Chloride Solution, 5000 µg/mL in Methanol 1 * 5 mL
Catalog # EHERGA09010500ME
Supplier: EHRENSTORFER
Frequently Bought Together
in Acetonitrile 1 * 1 mL
Supplier: EHRENSTORFER
pH buffers, for LAQUAtwin meters
Supplier: HORIBA
Supelco® Organic Reference Standard, 1-Hexene
Supplier: Merck
Carbomer Homopolymer, Reference Standard
Supplier: USP
Ofurace certified 1 * 0,1 g
Supplier: EHRENSTORFER
Supelco® Organic Reference Standard, Hexanal
Supplier: Merck
Chitosan, Reference Standard
Supplier: USP
Supelco® Organic Reference Standard, α-Tocotrienol
Supplier: Merck
POLY(VINYL ALCOHOL) 1 * 100 mg
Supplier: EDQM
Hexachlorobenzene 13C6 100 ng/µl 1 * 1 mL
Supplier: EHRENSTORFER
Methylsulfonylmethane, Reference Standard
Supplier: USP
IODFENPHOS 1 * 50 mg
Supplier: Merck
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
Perfluoropropanesulfonic Acid Sodium
Supplier: EHRENSTORFER
Supelco® Organic Reference Standard, Protein Standard Set
Supplier: Merck
V-SOLV ICP SOLVENT, 1 GAL IN SOLVENT 1 * 1 items
Supplier: LGC Standards PROMOCHEM