FERRI CHLORIDE REAGENT (50 AMPOULES OF 0.5 ML) 1 * 25 mL
Catalog # DIFC261190
Supplier: DIFCO BD
Frequently Bought Together
Pasteur pipettes, disposable, Jumbo Pastettes®
Supplier: Alpha Laboratories
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR®, Breatheable Rayon Film Seals for Biological Cultures
Supplier: VWR Collection
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
Cryoboxes, 133×133 mm
Supplier: RATIOLAB
Serological pipettes, Stripette™
Supplier: Corning
VWR®, Microcentrifuge Tubes with Socket Screw Caps
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Scoops
Supplier: VITLAB
Petri dishes, Corning® Gosselin™
Supplier: Corning
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Sample containers, snap-seal, Corning®
Supplier: Corning
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Storage rack for bags, BagRack® <i>Slide</i>
Supplier: INTERSCIENCE