Order Entry
Netherlands
ContactUsLinkComponent
An ELISA kit for the detection of Obestatin (Porcine). This uses Competitive ELISA, Coated with Antigen and has a sensitivity of 9.3 75pg/ml. 1 * 1 KIT
  GENOIT6382
An ELISA kit for the detection of Obestatin (Porcine). This uses Competitive ELISA, Coated with Antigen and has a sensitivity of 9.3 75pg/ml. 1 * 1 KIT
  GENOIT6382
Frequently Bought Together