Order Entry
Netherlands
ContactUsLinkComponent
Human CD30, Fc Tag (CD0-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession NP001234.2). 1 * 100 µG
Catalog # ACRBCD0-H5250100UG
Supplier:  ACRO Biosystems
Human CD30, Fc Tag (CD0-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession NP001234.2). 1 * 100 µG
Catalog # ACRBCD0-H5250100UG
Supplier:  ACRO Biosystems
Frequently Bought Together