Order Entry
Netherlands
ContactUsLinkComponent
Alexa Fluor® 488 Phalloidin allows researchers to fluorescently stain the cytoskeleton through the binding of phalloidin to F-actin. This product is intended for use on fixed and permeabilized samples due to the toxicity associated with phalloidin. A 1 * 1 items
Catalog # CSTA8878S
Supplier:  Cell Signaling
Alexa Fluor® 488 Phalloidin allows researchers to fluorescently stain the cytoskeleton through the binding of phalloidin to F-actin. This product is intended for use on fixed and permeabilized samples due to the toxicity associated with phalloidin. A 1 * 1 items
Catalog # CSTA8878S
Supplier:  Cell Signaling
Frequently Bought Together