Order Entry
Netherlands
ContactUsLinkComponent
Oxoid Ceftazidime/Avibactam Disk - (10/4) x 1 Packaging 5 Cartridges/Pk. Quantity 50 Disks/Cartridge Concentration 10/4µg 1 * 5 items
Catalog # OXOICT1948B
Oxoid Ceftazidime/Avibactam Disk - (10/4) x 1 Packaging 5 Cartridges/Pk. Quantity 50 Disks/Cartridge Concentration 10/4µg 1 * 5 items
Catalog # OXOICT1948B
Frequently Bought Together