Order Entry
Netherlands
ContactUsLinkComponent
Stericlin® sterilisation wrap interleaved, alternately stacked sheets, SSMMS 43g/m² light blue and non-woven 57g/m² green, 120x120cm , DIN EN ISO 11607 1 * 120 items
Catalog # 129-1340
Stericlin® sterilisation wrap interleaved, alternately stacked sheets, SSMMS 43g/m² light blue and non-woven 57g/m² green, 120x120cm , DIN EN ISO 11607 1 * 120 items
Catalog # 129-1340
Frequently Bought Together