Order Entry
Netherlands
ContactUsLinkComponent
Stericlin® sterilisation double wrap, 2 sheets sealed at the edges, outer sheet SSMMS 55g/m² dark blue, inner sheet non-woven 67g/m² green, 90x90cm, DIN EN ISO 11607 1 * 100 items
Catalog # 129-1334
Stericlin® sterilisation double wrap, 2 sheets sealed at the edges, outer sheet SSMMS 55g/m² dark blue, inner sheet non-woven 67g/m² green, 90x90cm, DIN EN ISO 11607 1 * 100 items
Catalog # 129-1334
Frequently Bought Together