Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Stericlin® thermal transfer film AXR® 8, for plastic substrates, width 60mm x length 74m x core diameter 12.7mm 1 * 5 Roll
Catalog # VEPA3FSGZ815480
Supplier:
VP Medical Packaging
Expand image
Stericlin® thermal transfer film AXR® 8, for plastic substrates, width 60mm x length 74m x core diameter 12.7mm 1 * 5 Roll
Catalog # VEPA3FSGZ815480
Supplier:
VP Medical Packaging
Frequently Bought Together
VWR® Nitrile Extra Light, Nitrile Gloves
Supplier: VWR Collection
Nitrile gloves, TouchNTuff® 92-600 / 92-605
Supplier: Ansell
Nitrile gloves, Kimtech™ Purple Nitrile™ Xtra™
Supplier: KIMBERLY CLARK
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
Neoprene gloves, Microflex® NeoTouch® 25-101 / 25-201
Supplier: Ansell
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Laboratory Markers
Supplier: VWR Collection
Lead-free steam indicator tapes, Comply™
Supplier: 3M MEDICAL
VWR® Labels, Hazardous Substance, GHS Labels
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR® Stockholm, Safety Spectacles
Supplier: VWR Collection
VWR®, Latex Gloves
Supplier: VWR Collection