Order Entry
Netherlands
ContactUsLinkComponent
Stericlin® barcode-labels, on roll, for thermal transfer printers, paper, double self-adhesive (sandwich), with 3 STEAM indicators, label size 102x56mm split in T shape, roll width 105mm x core 76mm, pack of 10 rolls of 1000 labels 1 * 10.000 items
Catalog # VEPA3FSKS675165
Stericlin® barcode-labels, on roll, for thermal transfer printers, paper, double self-adhesive (sandwich), with 3 STEAM indicators, label size 102x56mm split in T shape, roll width 105mm x core 76mm, pack of 10 rolls of 1000 labels 1 * 10.000 items
Catalog # VEPA3FSKS675165
Frequently Bought Together