Order Entry
Netherlands
ContactUsLinkComponent
Stericlin® sterilisation wrap sheet, SSMMS 43g/m², one side light blue, one side violet, 140x140cm, DIN EN ISO 11607 1 * 120 items
Catalog # 129-1365
Stericlin® sterilisation wrap sheet, SSMMS 43g/m², one side light blue, one side violet, 140x140cm, DIN EN ISO 11607 1 * 120 items
Catalog # 129-1365
Frequently Bought Together