Order Entry
Netherlands
ContactUsLinkComponent
Stericlin® sterilisation wrap sheet, SSMMS 43g/m², one side light blue, one side violet, 100x100cm, DIN EN ISO 11607 1 * 200 items
Catalog # 129-1362
Stericlin® sterilisation wrap sheet, SSMMS 43g/m², one side light blue, one side violet, 100x100cm, DIN EN ISO 11607 1 * 200 items
Catalog # 129-1362
Frequently Bought Together