m6A RNA Methylation Assay Kit (Fluorometric) 1 * 96 Tests
Catalog # ABCAAB233491-96
Supplier: Abcam
Frequently Bought Together
Media bottles, square, sterile, with screw cap, Nalgene®
Supplier: Thermo Fisher Scientific
Sampling spoons with long handle, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Cryo.S™ tubes, 1 and 2 ml
Supplier: Greiner Bio-One
VWR® Plugs for Drosophila Vials and Bottles
Supplier: VWR Collection
VWR® Media Bottles, Square, PETG
Supplier: VWR Collection
Microplates, 384-well, non binding
Supplier: Greiner Bio-One
Filter plates, 24-well, AcroPrep™
Supplier: Cytiva (Formerly Pall Lab)
Stoppers, press down, VacuCap®
Supplier: Simport Scientific
Isofreeze® racks with temperature indicator
Supplier: SARSTEDT
PCR microtubes with caps, 0.2 ml and 0.5 ml, Amplitube™
Supplier: Simport Scientific
Anti-YTHDF2 Rabbit Monoclonal Antibody [clone: EPR20318]
Supplier: Abcam
96-Well clear polystyrene microplates, Corning®
Supplier: Corning
Collagen I-coated microplates, Corning® BioCoat®
Supplier: Corning
FalconRegistered Mark Serological Pipettes, 25 mL 1 * 200 items
Supplier: STEMCELL Technologies