PTFE-tubing, natural inner-0: 10,00 mm +/- 0,20 mm outer-0: 12,00 mm +1- 0,20 mm wall: 1,00 mm in coils of 100M Material: PTFE acc to FDA and USP class VI 1 * 50 m
Catalog # VERNA210100010010A
Supplier: Saint Gobain Life Sciences
Frequently Bought Together