(R)-(-)-2-Hydroxyhex-5-ene
Bottle, 250 mg
Catalog # APOSOR304052-250MG
Supplier: Apollo Scientific
(R)-(-)-2-Hydroxyhex-5-ene
Catalog # APOSOR304052-250MG
Supplier: Apollo Scientific
CAS Number: 17397-29-4
Jump to:
Product Details & Documents
- CAS Number:17397-29-4
- Formula:C₆H₁₂O
- MDL Number:MFCD03427201
- Molecular Weight:100.159 g/mol
- Boiling Pt:159 °C
- Density:0.828 g/cm³ (25 °C)
- Flash Pt:43 °C
- Storage Temperature:Ambient
- UN:1987
- ADR:3,III
Specifications
Specifications
- Pack type:Bottle
- Pk:250 mg



