Order Entry
Netherlands
Orders LinkContactUsLinkComponent
(R)-(-)-2-Hydroxyhex-5-ene

(R)-(-)-2-Hydroxyhex-5-ene

Bottle, 1 g
Catalog # APOSOR304052-1G
Flammable
Supplier:   Apollo Scientific
(R)-(-)-2-Hydroxyhex-5-ene
Catalog # APOSOR304052-1G
Supplier:  Apollo Scientific
CAS Number:  17397-29-4
(R)-(-)-2-Hydroxyhex-5-ene

Bottle, 1 g

CAS Number:  17397-29-4
Price
167,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:17397-29-4
  • Formula:C₆H₁₂O
  • MDL Number:MFCD03427201
  • Molecular Weight:100.159 g/mol
  • Boiling Pt:159 °C
  • Density:0.828 g/cm³ (25 °C)
  • Flash Pt:43 °C
  • Storage Temperature:Ambient
  • UN:1987
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications