SILVER STEARATE 1 * 5 g
Catalog # MOLE90005152-5G
Supplier: Molekula
Frequently Bought Together
Ethanol eurodenatured ≥80%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
4-Methyl-2-pentanone ≥97%, TECHNICAL
Supplier: VWR Chemicals
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Methyl ethyl ketone ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Visitor Coats, PP, Disposable, Elasticated Wrist
Supplier: VWR Collection
Dimethyl sulphoxide, dehydrated (max. 0.01% H₂O) ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanolamine, EMSURE® for analysis, Supelco®
Supplier: Merck
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers) ≥98.5% (isomers plus ethylbenzene) ACS
Supplier: Thermo Fisher Scientific
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals