Order Entry
Netherlands
ContactUsLinkComponent
Ground Joint-GL Tube Fittings, PTFE. For transition from ground joints to GL threads. Cone size NS 29/32. Bore Ø 20,0 mm. Thread GL 45. 1 * 1 items
Catalog # BOHLD570-45
Supplier:  Bohlender
undefined
Ground Joint-GL Tube Fittings, PTFE. For transition from ground joints to GL threads. Cone size NS 29/32. Bore Ø 20,0 mm. Thread GL 45. 1 * 1 items
Catalog # BOHLD570-45
Supplier:  Bohlender
Frequently Bought Together