Order Entry
Netherlands
ContactUsLinkComponent
KingFisher 96 Accessory Kit A for 4 x 96 NucleoMag Tissue / Trace / Virus preps using a King Fisher 96 platform - Square-well Blocks , Deep-well Tip Combs, Elution Plates 1 * 24 SET
Catalog # MANA744950
Supplier:  MACHEREY-NAGEL
undefined
KingFisher 96 Accessory Kit A for 4 x 96 NucleoMag Tissue / Trace / Virus preps using a King Fisher 96 platform - Square-well Blocks , Deep-well Tip Combs, Elution Plates 1 * 24 SET
Catalog # MANA744950
Supplier:  MACHEREY-NAGEL
Frequently Bought Together