NucleoSpin totalRNA FFPE (10) 10 preps for the purification of small and large RNA from FFPE samples - NucleoSpin RNA Columns, Colle ction Tubes, Paraffin Dissolver, buffers, Proteinase K, RNase-free rDNase EQ Excepted Quantities or __UN 1993 Flam 1 * 10 items
Catalog # MANA740982.10
Supplier: MACHEREY-NAGEL
Frequently Bought Together