Decrosslink Buffer D-Link (30 mL) Decrosslink Buffer D-Link to be used with NucleoSpin FFPE kits bottle of 30 mL 1 * 30 mL
MANA740979.30
Decrosslink Buffer D-Link (30 mL) Decrosslink Buffer D-Link to be used with NucleoSpin FFPE kits bottle of 30 mL 1 * 30 mL
MANA740979.30
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection