Order Entry
Netherlands
ContactUsLinkComponent
NucleoSpin Blood (240) 240 preps for the isolation of genomic DNA from blood Buffers, Proteinase K, NucleoSpin Blood Columns __UN 1993 Flammable liquid, n.o.s. (2-propanol mixture) 3 III 0.250 L ADR/GGVSEB F1 (E) LQ 1 * 240 items
Catalog # MANA740951.240C
Supplier:  MACHEREY-NAGEL
undefined
NucleoSpin Blood (240) 240 preps for the isolation of genomic DNA from blood Buffers, Proteinase K, NucleoSpin Blood Columns __UN 1993 Flammable liquid, n.o.s. (2-propanol mixture) 3 III 0.250 L ADR/GGVSEB F1 (E) LQ 1 * 240 items
Catalog # MANA740951.240C
Supplier:  MACHEREY-NAGEL
Frequently Bought Together