NucleoSpin RNA/DNA Buffer Set buffer set for separated isolation of RNA and DNA from one lysate, only applicable with NucleoSpin RNA , NucleoSpin RNA XS, NucleoSpin miRNA, NS RNA Blood, NucleoSpin RNA Plant, or NucleoSpin RNA/Protein kits, sufficient 1 * 1 SET
Catalog # MANA740944
Supplier: MACHEREY-NAGEL
Frequently Bought Together