NucleoSpin 96 DNA Plasma Core Kit(4 x 96) 4 x 96 preps for the isolation of cell-free DNA from blood plasma - NucleoSpin 96 Plasma B inding Plates, Buffers, Proteinase K __UN 1993 Flammable liquid, n.o.s. (ethanol, 2-propanol mixture) 1 * 384 items
MANA740874.4
Frequently Bought Together