Order Entry
Netherlands
ContactUsLinkComponent
NucleoSpin 96 DNA Plasma Core Kit(1 x 96) 1 x 96 preps for the isolation of cell-free DNA from blood plasma - NucleoSpin 96 Plasma B inding Plate, Buffers, Proteinase K __UN 1993 Flammable liquid, n.o.s. (ethanol, 2-propanol mixture) 1 * 96 items
Catalog # MANA740874.1
Supplier:  MACHEREY-NAGEL
undefined
NucleoSpin 96 DNA Plasma Core Kit(1 x 96) 1 x 96 preps for the isolation of cell-free DNA from blood plasma - NucleoSpin 96 Plasma B inding Plate, Buffers, Proteinase K __UN 1993 Flammable liquid, n.o.s. (ethanol, 2-propanol mixture) 1 * 96 items
Catalog # MANA740874.1
Supplier:  MACHEREY-NAGEL
Frequently Bought Together