NucleoSpin 96 DNA Plasma (1x96) 1 x 96 preps for the isolation of cell-free DNA from blood plasma - NucleoSpin 96 Plasma Binding Pla te, Buffers, Proteinase K , 24-Square-well Blocks, Round-well Block, NucleoSpin 96 Plasma Binding Plate __UN 1993 F 1 * 96 items
Catalog # MANA740873.1
Supplier: MACHEREY-NAGEL
Frequently Bought Together