Extraction thimbles MN 645 Format: 10 x 50 mm (Inner diameter x lenght) external diameter 14 mm pack of 25 1 * 25 items
MANA645089.1
Extraction thimbles MN 645 Format: 10 x 50 mm (Inner diameter x lenght) external diameter 14 mm pack of 25 1 * 25 items
MANA645089.1
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection