Order Entry
Netherlands
ContactUsLinkComponent
Buffer NT2 (100 mL) Binding Buffer NT2 1 bottle of 100 mL __UN 3211 Perchlorates, inorganic, aqueous solution, n.o.s. (Sod ium perchlorate) 5.1 III 0.100 L ADR/GGVSEB O1 (E) LQ PAX 551 2.5 L CAO 555 30 L Q: 0.040 1 * 100 mL
Catalog # MANA740597
Supplier:  MACHEREY-NAGEL
undefined
Buffer NT2 (100 mL) Binding Buffer NT2 1 bottle of 100 mL __UN 3211 Perchlorates, inorganic, aqueous solution, n.o.s. (Sod ium perchlorate) 5.1 III 0.100 L ADR/GGVSEB O1 (E) LQ PAX 551 2.5 L CAO 555 30 L Q: 0.040 1 * 100 mL
Catalog # MANA740597
Supplier:  MACHEREY-NAGEL
Frequently Bought Together