Order Entry
Netherlands
ContactUsLinkComponent
NucleoTrap (100) 100 preps for gel extraction - NucleoTrap Suspension, buffers pack of 100 __UN 3211 Perchlorates, inorganic, aque ous solution, n.o.s. 5.1 III 0.160 L ADR/GGVSEB O1 (E) LQ PAX 551 2.5 1 * 100 items
Catalog # MANA740584
Leverancier:  MACHEREY-NAGEL
undefined
NucleoTrap (100) 100 preps for gel extraction - NucleoTrap Suspension, buffers pack of 100 __UN 3211 Perchlorates, inorganic, aque ous solution, n.o.s. 5.1 III 0.160 L ADR/GGVSEB O1 (E) LQ PAX 551 2.5 1 * 100 items
Catalog # MANA740584
Leverancier:  MACHEREY-NAGEL
Frequently Bought Together